Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R2 SIRNA Assays ›
Gene Symbol:
PPP6R2
Gene Name:
protein phosphatase 6 regulatory subunit 2
Gene Aliases:
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location:
Chr.22: 50343317 - 50445090 on Build GRCh38
UniGene ID:
Hs.449098
Species:
Human
siRNA ID
HSS190541
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242898.1
NP_001229827.1
7
1059
NM_001242899.1
NP_001229828.1
7
1059
NM_001242900.1
NP_001229829.1
7
1059
NM_014678.4
NP_055493.2
7
1059
XM_006724431.2
XP_006724494.1
8
1170
XM_006724434.1
XP_006724497.1
9
1319
XM_006724435.2
XP_006724498.1
7
1044
XM_011530720.2
XP_011529022.1
6
744
XM_011530721.2
XP_011529023.1
6
744
XM_011530722.2
XP_011529024.1
6
744
XM_011530723.2
XP_011529025.1
6
744
XM_011530724.2
XP_011529026.1
6
744
XM_011530726.2
XP_011529028.1
6
744
XM_011530727.2
XP_011529029.1
6
744
XM_011530728.2
XP_011529030.1
6
744
XM_011530729.2
XP_011529031.1
6
744
XM_011530730.2
XP_011529032.1
6
744
XM_011530731.1
XP_011529033.1
6
744
XM_011530732.2
XP_011529034.1
9
1220
XM_011530734.2
XP_011529036.1
8
1094
XM_011530735.2
XP_011529037.1
7
1044
XM_011530736.2
XP_011529038.1
8
1170
XM_011530737.2
XP_011529039.1
9
1319
XM_011530738.2
XP_011529040.1
8
1122
XM_011530739.2
XP_011529041.1
6
784
XM_011530740.2
XP_011529042.1
8
1192
XM_017029116.1
XP_016884605.1
6
744
XM_017029117.1
XP_016884606.1
10
1369
XM_017029118.1
XP_016884607.1
10
1321
XM_017029119.1
XP_016884608.1
9
1243
XM_017029120.1
XP_016884609.1
9
1220
XM_017029121.1
XP_016884610.1
7
1044
XM_017029122.1
XP_016884611.1
8
1122
XM_017029123.1
XP_016884612.1
7
1044
XM_017029124.1
XP_016884613.1
8
1170
XM_017029125.1
XP_016884614.1
7
1044
XM_017029126.1
XP_016884615.1
7
1044
XM_017029127.1
XP_016884616.1
8
1122
XM_017029128.1
XP_016884617.1
8
1170
XM_017029129.1
XP_016884618.1
7
1044
XM_017029130.1
XP_016884619.1
7
1044
XM_017029131.1
XP_016884620.1
8
1170
XM_017029132.1
XP_016884621.1
8
1122
XM_017029133.1
XP_016884622.1
7
998
XM_017029134.1
XP_016884623.1
7
998
XM_017029135.1
XP_016884624.1
8
1048
XM_017029136.1
XP_016884625.1
7
998
XM_017029137.1
XP_016884626.1
7
998
XM_017029138.1
XP_016884627.1
7
998
GenBank mRNA
AB014585.1
-
7
1031
BC000976.4
-
8
1048
2900437
BC006568.2
-
7
1045
3532211
BC032664.1
-
7
1056
5553142
BC041698.1
-
7
1026
5732101
BC052995.2
-
7
965
6162781
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0