Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014705.3 | NP_055520.3 | 12, 13 | 1338 | ||
XM_006716188.2 | XP_006716251.1 | 12, 13 | 1325 | |||
XM_006716189.2 | XP_006716252.1 | 12, 13 | 1325 | |||
XM_017012819.1 | XP_016868308.1 | 12, 13 | 1698 | |||
XM_017012820.1 | XP_016868309.1 | 12, 13 | 1261 | |||
XM_017012821.1 | XP_016868310.1 | 12, 13 | 1252 | |||
XM_017012822.1 | XP_016868311.1 | 12, 13 | 1843 | |||
XM_017012823.1 | XP_016868312.1 | 12, 13 | 1252 | |||
XM_017012824.1 | XP_016868313.1 | 12, 13 | 1252 | |||
XM_017012825.1 | XP_016868314.1 | 12, 13 | 1252 | |||
GenBank mRNA | AY233380.1 | - | 12, 13 | 1069 | ||
BC117688.2 | - | 12, 13 | 1069 | 40072000 | ||
BC117689.1 | - | 12, 13 | 1069 | 40072006 | ||
BC117694.1 | - | 12, 13 | 1069 | 40072219 |