Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
siRNA ID
HSS190582
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
9
1212
NM_001204145.2
NP_001191074.1
7
904
NM_001204146.2
NP_001191075.1
7
1082
NM_001204147.2
NP_001191076.1
6
864
NM_001204148.2
NP_001191077.1
7
1011
NM_001321868.1
NP_001308797.1
8
1152
NM_001321869.1
NP_001308798.1
9
1293
NM_001321870.1
NP_001308799.1
9
1284
NM_001321871.1
NP_001308800.1
8
1167
NM_001321872.1
NP_001308801.1
8
1161
NM_001321873.1
NP_001308802.1
8
1155
NM_001321874.1
NP_001308803.1
8
1214
NM_001321875.1
NP_001308804.1
8
1220
NM_001321876.1
NP_001308805.1
8
1208
NM_001321877.1
NP_001308806.1
7
1082
NM_001321878.1
NP_001308807.1
8
1205
NM_001321879.1
NP_001308808.1
7
1076
NM_001321884.1
NP_001308813.1
8
1208
NM_001321885.1
NP_001308814.1
7
1097
NM_001321886.1
NP_001308815.1
9
1264
NM_001321887.1
NP_001308816.1
9
1098
NM_001321888.1
NP_001308817.1
7
898
NM_001321889.1
NP_001308818.1
8
1135
NM_001321890.1
NP_001308819.1
8
1199
NM_001321891.1
NP_001308820.1
7
1073
NM_001321893.1
NP_001308822.1
8
970
NM_001321894.1
NP_001308823.1
8
979
NM_001321895.1
NP_001308824.1
7
913
NM_001321896.1
NP_001308825.1
9
1383
NM_001321897.1
NP_001308826.1
8
979
NM_001321898.1
NP_001308827.1
7
928
NM_001321899.1
NP_001308828.1
9
1111
NM_001321900.1
NP_001308829.1
8
1205
NM_001321901.1
NP_001308830.1
8
1039
NM_001321902.1
NP_001308831.1
8
1045
NM_014707.3
NP_055522.1
8
1036
NM_058176.2
NP_478056.1
7
891
NM_178423.2
NP_848510.1
8
1036
NM_178425.3
NP_848512.1
7
900
XM_011515629.2
XP_011513931.1
8
994
XM_011515631.2
XP_011513933.1
9
1308
XM_011515634.2
XP_011513936.1
7
862
XM_011515635.2
XP_011513937.1
9
1860
XM_011515636.2
XP_011513938.1
8
1581
XM_011515637.2
XP_011513939.1
8
1005
XM_011515638.2
XP_011513940.1
8
1575
XM_011515639.2
XP_011513941.1
8
1772
XM_011515641.2
XP_011513943.1
9
1857
XM_011515642.2
XP_011513944.1
7
2586
XM_011515644.2
XP_011513946.1
9
1832
XM_011515645.2
XP_011513947.1
7
2571
XM_011515646.2
XP_011513948.1
7
2562
XM_011515648.2
XP_011513950.1
8
1590
XM_011515649.2
XP_011513951.1
8
1794
XM_011515651.1
XP_011513953.1
7
828
XM_017012826.1
XP_016868315.1
8
1566
XM_017012827.1
XP_016868316.1
9
1649
XM_017012828.1
XP_016868317.1
7
1642
XM_017012829.1
XP_016868318.1
8
969
XM_017012830.1
XP_016868319.1
8
960
XM_017012831.1
XP_016868320.1
7
837
XM_017012832.1
XP_016868321.1
8
969
GenBank mRNA
AB018287.1
-
8
1000
AF124924.1
-
5
558
AJ459808.1
-
8
1000
AK097178.1
-
7
862
AK296214.1
-
7
814
AK297404.1
-
7
858
AK304298.1
-
9
1205
AK304343.1
-
7
931
AK304410.1
-
6
827
AK316109.1
-
7
1033
AY032737.1
-
7
850
AY032738.1
-
7
850
AY197371.1
-
7
859
BC111735.1
-
6
726
BC150328.1
-
8
1011
8860495
BC152405.1
-
8
1000
8862508
CR749550.1
-
7
967
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0