Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014753.3 | NP_055568.3 | 10 | 2168 | ||
XM_005271846.3 | XP_005271903.1 | 10 | 2043 | |||
XM_005271848.3 | XP_005271905.1 | 10 | 1899 | |||
XM_005271849.3 | XP_005271906.1 | 10 | 1899 | |||
XM_011540402.2 | XP_011538704.1 | 10 | 1899 | |||
XM_011540403.2 | XP_011538705.1 | 2 | 702 | |||
GenBank mRNA | AK308420.1 | - | 10 | 1875 | ||
BC043345.1 | - | 10 | 1863 | 6062924 | ||
BC150252.1 | - | 9 | 1837 | 9007274 | ||
D80009.1 | - | 9 | 1837 |