Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290474.1 | NP_001277403.1 | 13 | 2155 | ||
NM_014807.4 | NP_055622.3 | 13 | 2158 | |||
XM_006718948.3 | XP_006719011.1 | 12 | 2336 | |||
XM_011543076.2 | XP_011541378.1 | 13 | 2654 | |||
XM_017018625.1 | XP_016874114.1 | 12 | 2333 | |||
XM_017018626.1 | XP_016874115.1 | 12 | 2246 | |||
XM_017018627.1 | XP_016874116.1 | 12 | 2243 | |||
XM_017018628.1 | XP_016874117.1 | 13 | 1580 | |||
GenBank mRNA | AB006623.2 | - | 13 | 1620 | ||
AB094094.1 | - | 13 | 2126 | |||
AK025223.1 | - | 13 | 2126 | |||
BC003598.1 | - | 5 | 640 | |||
BC010071.2 | - | 7 | 848 | |||
BC021254.2 | - | 7 | 848 | |||
BC022219.1 | - | 13 | 2021 | 3905001 | ||
BC033083.1 | - | 7 | 848 | |||
BC052246.1 | - | 5 | 628 | |||
BC075832.1 | - | 7 | 848 | |||
BX537572.1 | - | 7 | 3030 |