Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011553.3 | NP_001011553.2 | 1 | 262 | ||
NM_001242956.1 | NP_001229885.1 | 1 | 262 | |||
NM_001788.5 | NP_001779.3 | 1 | 262 | |||
XM_006715806.3 | XP_006715869.1 | 1 | 210 | |||
XM_011515656.2 | XP_011513958.1 | 1 | 210 | |||
XM_011515661.2 | XP_011513963.1 | 1 | 210 | |||
XM_017012855.1 | XP_016868344.1 | 1 | 203 | |||
XM_017012856.1 | XP_016868345.1 | 1 | 208 | |||
XM_017012857.1 | XP_016868346.1 | 1 | 208 | |||
XM_017012858.1 | XP_016868347.1 | 1 | 30 | |||
XM_017012859.1 | XP_016868348.1 | 1 | 30 | |||
XM_017012860.1 | XP_016868349.1 | 1 | 30 | |||
XM_017012861.1 | XP_016868350.1 | 1 | 30 | |||
XM_017012862.1 | XP_016868351.1 | 1 | 30 | |||
XM_017012863.1 | XP_016868352.1 | 1 | 30 | |||
GenBank mRNA | AB073389.1 | - | 1 | 29 | ||
AK223225.1 | - | 1 | 29 | |||
AK223241.1 | - | 1 | 29 | |||
AK290545.1 | - | 1 | 103 | |||
AK297883.1 | - | 1 | 29 | |||
AK298453.1 | - | 1 | 29 | |||
BC025987.2 | - | 1 | 29 | 4823810 | ||
BC093640.1 | - | 1 | 28 | 7939485 | ||
BC093642.1 | - | 1 | 28 | 7939487 | ||
BC107718.1 | - | 1 | 210 | |||
BI669760.1 | - | 1 | 197 | |||
BP280069.1 | - | 1 | 233 | |||
BQ229116.1 | - | 1 | 193 | |||
BX571755.1 | - | 1 | 30 | |||
DC414394.1 | - | 1 | 265 | |||
DQ232879.1 | - | 1 | 13 | |||
S72008.1 | - | 1 | 29 |