Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114091.2 | NP_001107563.1 | 14 | 1908 | ||
NM_001256.4 | NP_001247.3 | 14 | 1890 | |||
NM_001293089.1 | NP_001280018.1 | 14 | 1887 | |||
NM_001293091.1 | NP_001280020.1 | 13 | 1742 | |||
XM_011525546.2 | XP_011523848.1 | 14 | 1892 | |||
XM_011525547.2 | XP_011523849.1 | 14 | 1895 | |||
XM_011525548.2 | XP_011523850.1 | 13 | 1780 | |||
XM_011525549.2 | XP_011523851.1 | 13 | 1903 | |||
XM_011525550.2 | XP_011523852.1 | 12 | 1590 | |||
XM_017025481.1 | XP_016880970.1 | 14 | 1892 | |||
XM_017025482.1 | XP_016880971.1 | 14 | 1877 | |||
XM_017025483.1 | XP_016880972.1 | 14 | 1874 | |||
XM_017025484.1 | XP_016880973.1 | 13 | 1755 | |||
XM_017025485.1 | XP_016880974.1 | 12 | 1633 | |||
XM_017025486.1 | XP_016880975.1 | 12 | 1550 | |||
XM_017025487.1 | XP_016880976.1 | 13 | 1761 | |||
XM_017025488.1 | XP_016880977.1 | 14 | 1857 | |||
GenBank mRNA | AK296881.1 | - | 13 | 1682 | ||
BC011656.2 | - | 14 | 1772 | 4301175 | ||
S78234.1 | - | 14 | 1850 | |||
U00001.1 | - | 14 | 1805 |