Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175305.4 | NP_780514.1 | 3 | 531 | ||
GenBank mRNA | AK033594.1 | - | 3 | 532 | ||
BC061228.1 | - | 4 | 571 | 30314668 |