Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089546.1 | - | 6 | 1373 | ||
AK155267.1 | - | 6 | 1373 | |||
AK164544.1 | - | 6 | 1321 | |||
BC026718.1 | - | 1 | 211 | |||
BC034249.1 | - | 7 | 1327 | 3675490 | ||
CX220787.1 | - | 4 | 427 |