Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199446.1 | NP_955517.1 | 29 | 2927 | ||
GenBank mRNA | AK078940.1 | - | 19 | 1781 | ||
AK137529.1 | - | 3 | 262 | |||
AK153136.1 | - | 7 | 639 | |||
BC053105.1 | - | 29 | 2927 | 6409425 |