Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam192a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028221.4 | NP_082497.2 | 6 | 747 | ||
GenBank mRNA | AK010053.1 | - | 6 | 712 | ||
AK033988.1 | - | 6 | 678 | |||
AK050704.1 | - | 6 | 682 | |||
AK151603.1 | - | 6 | 701 | |||
AK213161.1 | - | 2 | 100 | |||
BC016088.1 | - | 5 | 487 | 4503452 | ||
BC023158.1 | - | 6 | 586 | 4946201 | ||
BC025472.1 | - | 6 | 723 | 5353148 | ||
BC027060.1 | - | 6 | 709 | 5358022 | ||
BC027818.1 | - | 5 | 515 | 5320967 | ||
BC040818.1 | - | 700 | 4194531 |