Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081240.3 | NP_001074709.1 | 3 | 636 | ||
GenBank mRNA | AK045296.1 | - | 3 | 627 | ||
AK045315.1 | - | 3 | 626 | |||
AK154546.1 | - | 3 | 580 | |||
AK161023.1 | - | 3 | 580 | |||
AK205080.1 | - | 1 | 59 |