Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 3 | 535 | ||
GenBank mRNA | AK041905.1 | - | 3 | 404 | ||
AK089681.1 | - | 379 | ||||
AK090333.1 | - | 3 | 379 | |||
AK157968.1 | - | 3 | 388 | |||
AK170639.1 | - | 3 | 388 | |||
AK170923.1 | - | 3 | 358 | |||
AK172462.1 | - | 3 | 369 | |||
AK189240.1 | - | 1 | 29 | |||
AY166673.1 | - | 1 | 53 | |||
BC013467.1 | - | 2 | 86 | 4218025 | ||
BC075666.1 | - | 3 | 535 | 30605163 |