Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271347.1 | NP_001258276.1 | 7 | 1149 | ||
NM_001271348.1 | NP_001258277.1 | 7 | 1110 | |||
NM_001271349.1 | NP_001258278.1 | 6 | 1047 | |||
NM_134015.3 | NP_598776.1 | 8 | 1212 | |||
GenBank mRNA | AB093260.1 | - | 6 | 854 | ||
AK041532.1 | - | 7 | 942 | |||
AK149139.1 | - | 7 | 932 | |||
AK152181.1 | - | 7 | 872 | |||
AK160086.1 | - | 6 | 848 | |||
AK166226.1 | - | 7 | 807 | |||
AK168667.1 | - | 6 | 864 | |||
AK186752.1 | - | 1 | 54 | |||
AY038079.1 | - | 8 | 876 | |||
BC034261.1 | - | 8 | 892 | 4237375 |