Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038619.1 | NP_001033708.1 | 3, 4 | 597 | ||
NM_172646.2 | NP_766234.1 | 3, 4 | 601 | |||
GenBank mRNA | AK035951.1 | - | 3, 4 | 580 | ||
AK036199.1 | - | 3, 4 | 562 | |||
AK049724.1 | - | 3, 4 | 602 | |||
AK082698.1 | - | 3, 4 | 541 | |||
AK147678.1 | - | 3, 4 | 541 | |||
BC141143.1 | - | 3, 4 | 445 | 9055755 | ||
BC141144.1 | - | 3, 4 | 445 | 9055756 |