Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113514.1 | NP_001106986.1 | 4 | 413 | ||
NM_133721.2 | NP_598482.2 | 18 | 2234 | |||
GenBank mRNA | AJ344342.1 | - | 18 | 2237 | ||
AK083987.1 | - | 3 | 253 | |||
AK139690.1 | - | 4 | 401 | |||
AK153585.1 | - | 16 | 1696 | |||
BC049939.1 | - | 1 | 30 | |||
BC150824.1 | - | 18 | 2140 | 9087735 | ||
CF913701.1 | - | 4 | 413 |