Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253813.1 | NP_001240742.1 | 19 | 2602 | ||
NM_175344.4 | NP_780553.2 | 18 | 2539 | |||
GenBank mRNA | AK089300.1 | - | 19 | 2675 | ||
AK159411.1 | - | 19 | 2551 | |||
BC023745.1 | - | 1 | 28 | |||
BC060732.1 | - | 18 | 2539 | 6409332 |