Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178928.4 | NP_849259.2 | 7 | 628 | ||
GenBank mRNA | AK035227.1 | - | 7 | 628 | ||
AK053714.1 | - | 7 | 627 | |||
BC138923.1 | - | 7 | 589 | 8861945 |