Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172671.2 | NP_766259.2 | 4, 5 | 868 | ||
GenBank mRNA | AK044357.1 | - | 3, 4 | 176 | ||
AY255619.1 | - | 3, 4 | 208 | |||
BE987673.1 | - | 4, 5 | 405 |