Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148943.2 | NP_683745.2 | 19 | 2821 | ||
GenBank mRNA | AJ307017.1 | - | 19 | 2821 |