Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133985.2 | NP_598746.2 | 16, 17 | 1793 | ||
GenBank mRNA | AK031790.1 | - | 15, 16 | 1290 | ||
AK075837.1 | - | 15, 16 | 1301 | |||
AK129291.1 | - | 16, 17 | 1788 | |||
BC036148.1 | - | 7, 8 | 543 | |||
BC046453.1 | - | 17, 18 | 1817 | |||
BC060645.1 | - | 16, 17 | 1776 | 6852021 |