Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080755.2 | NP_001074224.1 | 9 | 2511 | ||
NM_001287139.1 | NP_001274068.1 | 9 | 2450 | |||
GenBank mRNA | AK034365.1 | - | 6 | 1060 | ||
AK131150.1 | - | 9 | 2449 | |||
AK132612.1 | - | 6 | 675 | |||
AK165882.1 | - | 8 | 931 | |||
BC060614.1 | - | 9 | 2440 |