Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163263.1 | NP_001156735.1 | 5 | 1048 | ||
NM_182999.2 | NP_892044.1 | 6 | 895 | |||
GenBank mRNA | AK043896.1 | - | 1 | 24 | ||
AK048862.1 | - | 5 | 1007 | |||
AK051278.1 | - | 5 | 1007 | |||
AK053783.1 | - | 6 | 883 | |||
AK082529.1 | - | 5 | 952 | |||
AK085436.1 | - | 5 | 1026 | |||
AK087860.1 | - | 5 | 976 | |||
AK163905.1 | - | 2 | 261 | |||
AK220492.1 | - | 5 | 964 | |||
BC004593.1 | - | 1 | 122 | |||
BC052482.1 | - | 6 | 834 | 6308280 |