Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173404.3 | NP_775580.1 | 3 | 2030 | ||
GenBank mRNA | AK033644.1 | - | 3 | 1669 | ||
AK035393.1 | - | 3 | 2092 | |||
AK037225.1 | - | 3 | 2011 | |||
AK040389.1 | - | 3 | 2031 | |||
BC117749.2 | - | 3 | 1643 | 40090489 |