Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 18 | 2311 | ||
GenBank mRNA | AK032769.1 | - | 18 | 2324 | ||
AK035673.1 | - | 18 | 2041 | |||
AK133794.1 | - | 9 | 1041 | |||
AK134667.1 | - | 18 | 2321 | |||
AK142716.1 | - | 18 | 2321 | |||
AK147763.1 | - | 18 | 2343 | |||
AK168381.1 | - | 18 | 2323 | |||
AK169052.1 | - | 18 | 2346 | |||
AK169631.1 | - | 18 | 2325 | |||
AK177572.1 | - | 2 | 97 | |||
BC031457.1 | - | 18 | 2318 | |||
BC032209.1 | - | 12 | 1466 | |||
BC035283.1 | - | 18 | 2311 | 4947455 |