Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025837.2 | NP_080113.1 | 3 | 218 | ||
GenBank mRNA | AF244360.1 | - | 3 | 200 | ||
AK003896.1 | - | 3 | 218 | |||
AK132777.1 | - | 3 | 219 | |||
AK217169.1 | - | 3 | 204 | |||
BC009068.1 | - | 3 | 196 | 2650791 | ||
BC051163.1 | - | 3 | 196 | 6466145 |