Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 1 | 92 | ||
GenBank mRNA | AF306557.1 | - | 1 | 96 | ||
AK036505.1 | - | 1 | 90 | |||
AK037629.1 | - | 1 | 90 | |||
AK047057.1 | - | 1 | 85 | |||
AK142165.1 | - | 1 | 57 | |||
AK154172.1 | - | 1 | 70 | |||
AK166022.1 | - | 1 | 74 | |||
AK166340.1 | - | 1 | 90 | |||
AK170290.1 | - | 1 | 89 | |||
AK171416.1 | - | 1 | 79 | |||
BC031409.1 | - | 1 | 35 | 4952710 |