Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 13 | 1903 | ||
GenBank mRNA | AF306557.1 | - | 13 | 1907 | ||
AK036505.1 | - | 14 | 2003 | |||
AK047057.1 | - | 13 | 1896 | |||
AK142165.1 | - | 13 | 1868 | |||
AK154172.1 | - | 13 | 1885 | |||
AK166022.1 | - | 13 | 1886 | |||
AK166340.1 | - | 13 | 1901 | |||
AK170290.1 | - | 13 | 1900 | |||
BC031409.1 | - | 13 | 1846 | 4952710 |