Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081412.2 | NP_001074881.1 | 17, 18 | 3187 | ||
GenBank mRNA | AK017213.1 | - | 2, 3 | 246 | ||
AK129482.1 | - | 14, 15 | 1377 | |||
BC048842.1 | - | 5, 6 | 431 | |||
BC060270.1 | - | 17, 18 | 2425 | |||
BC131682.1 | - | 18, 19 | 2592 | 40111128 |