Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163529.1 | NP_001157001.1 | 12 | 1575 | ||
NM_033615.2 | NP_291093.2 | 12 | 1575 | |||
GenBank mRNA | AB059632.1 | - | 12 | 1575 | ||
AB059633.1 | - | 12 | 1575 | |||
AF386072.1 | - | 8 | 942 | |||
AF472524.1 | - | 12 | 1307 | |||
AK048367.1 | - | 8 | 842 | |||
AK053825.1 | - | 12 | 1551 | |||
AY054984.1 | - | 12 | 1275 | |||
AY382193.1 | - | 12 | 1306 | |||
AY382194.1 | - | 12 | 1306 | |||
AY953137.1 | - | 7 | 850 | |||
BC138139.1 | - | 12 | 1298 | 8861158 |