Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007387.2 | NP_031413.1 | 9 | 903 | ||
GenBank mRNA | AK089401.1 | - | 9 | 894 | ||
AK128963.1 | - | 9 | 896 | |||
AK133756.1 | - | 9 | 899 | |||
AK154269.1 | - | 9 | 899 | |||
AK170658.1 | - | 9 | 903 | |||
BC013495.1 | - | 9 | 891 | |||
BC023343.1 | - | 9 | 885 | 5136781 | ||
X57199.1 | - | 9 | 874 |