Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053262.3 | NP_444492.1 | 3 | 432 | ||
GenBank mRNA | AF304306.1 | - | 3 | 428 | ||
AK049355.1 | - | 3 | 476 | |||
AK154803.1 | - | 3 | 447 | |||
AK155174.1 | - | 3 | 447 | |||
AK155202.1 | - | 3 | 452 | |||
AK155327.1 | - | 3 | 452 | |||
AK158327.1 | - | 3 | 447 | |||
AK170939.1 | - | 3 | 447 | |||
AY053570.1 | - | 3 | 446 | |||
BC038340.1 | - | 3 | 432 | 4458725 |