Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009619.4 | NP_033749.2 | 9, 10 | 913 | ||
GenBank mRNA | BC060975.1 | - | 9, 10 | 876 | ||
BC129920.1 | - | 9, 10 | 879 | |||
X64227.1 | - | 9, 10 | 923 |