Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009634.6 | NP_033764.2 | 13 | 1480 | ||
GenBank mRNA | AK049372.1 | - | 13 | 1452 | ||
AK051111.1 | - | 13 | 1452 | |||
AK152092.1 | - | 13 | 1452 | |||
AK166019.1 | - | 13 | 1448 | |||
AK168906.1 | - | 13 | 1445 | |||
AK170216.1 | - | 11 | 1515 | |||
AK171725.1 | - | 13 | 1461 | |||
BC020187.1 | - | 13 | 1456 | 4913637 | ||
U20225.1 | - | 13 | 1456 |