Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287172.1 | NP_001274101.1 | 6 | 889 | ||
NM_001287176.1 | NP_001274105.1 | 6 | 1012 | |||
NM_007431.3 | NP_031457.2 | 6 | 864 | |||
GenBank mRNA | AB473959.1 | - | 5 | 628 | ||
AK076459.1 | - | 6 | 799 | |||
AK161780.1 | - | 6 | 800 | |||
AK164002.1 | - | 6 | 857 | |||
AK166732.1 | - | 6 | 798 | |||
AK167023.1 | - | 6 | 799 | |||
AK167481.1 | - | 6 | 799 | |||
BC065175.1 | - | 6 | 787 | 6807509 | ||
J02980.1 | - | 6 | 803 | |||
X13409.1 | - | 6 | 747 |