Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256382.1 | NP_001243311.1 | 9, 10 | 2304 | ||
NM_001256383.1 | NP_001243312.1 | 9, 10 | 2304 | |||
NM_053271.2 | NP_444501.1 | 11, 12 | 2424 | |||
GenBank mRNA | AB021131.1 | - | 11, 12 | 2378 | ||
AK129206.1 | - | 6, 7 | 1390 | |||
BC118979.2 | - | 9, 10 | 2252 | 40098740 | ||
HM015529.1 | - | 9, 10 | 2227 |