Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291146.1 | NP_001278075.1 | 6 | 936 | ||
NM_001291147.1 | NP_001278076.1 | 6 | 680 | |||
NM_001291148.1 | NP_001278077.1 | 6 | 642 | |||
NM_033562.4 | NP_291040.1 | 6 | 645 | |||
GenBank mRNA | AF208064.1 | - | 6 | 584 | ||
AK048627.1 | - | 6 | 592 | |||
AK077659.1 | - | 6 | 596 | |||
AK082564.1 | - | 6 | 605 | |||
AK083632.1 | - | 6 | 584 | |||
AK083725.1 | - | 3 | 3348 | |||
AK151934.1 | - | 6 | 936 | |||
BC005682.1 | - | 6 | 584 | 3154448 | ||
BC037503.1 | - | 2 | 227 |