Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172506.2 | NP_766094.1 | 3 | 738 | ||
GenBank mRNA | AK028770.1 | - | 3 | 759 | ||
AK028889.1 | - | 3 | 739 | |||
AK043379.1 | - | 3 | 748 | |||
BC062892.1 | - | 3 | 718 | 6418202 | ||
BC078631.1 | - | 3 | 685 | 6515418 |