Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301211.1 | NP_001288140.1 | 14 | 1730 | ||
NM_009677.6 | NP_033807.2 | 15 | 1739 | |||
GenBank mRNA | AK028360.1 | - | 3 | 145 | ||
AK034198.1 | - | 12 | 968 | |||
AK036387.1 | - | 15 | 1682 | |||
AK145821.1 | - | 14 | 1661 | |||
BC006691.1 | - | 14 | 1406 | |||
BC052703.1 | - | 15 | 1612 | 5698075 | ||
BC054535.1 | - | 15 | 1612 | 5698075 | ||
X54424.1 | - | 14 | 1652 |