Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301211.1 | NP_001288140.1 | 18 | 2129 | ||
NM_009677.6 | NP_033807.2 | 19 | 2138 | |||
GenBank mRNA | AK028360.1 | - | 7 | 544 | ||
AK034198.1 | - | 16 | 1367 | |||
AK036387.1 | - | 19 | 2081 | |||
AK145821.1 | - | 18 | 2060 | |||
BC006691.1 | - | 18 | 1805 | |||
BC052703.1 | - | 19 | 2011 | 5698075 | ||
BC054535.1 | - | 19 | 2011 | 5698075 | ||
X54424.1 | - | 18 | 2051 |