Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009703.2 | NP_033833.1 | 10 | 1060 | ||
GenBank mRNA | AK034861.1 | - | 10 | 1055 | ||
AK085823.1 | - | 10 | 1026 | |||
AK135358.1 | - | 10 | 1026 | |||
AK163567.1 | - | 10 | 1029 | |||
AK169757.1 | - | 10 | 1042 | |||
BC004757.1 | - | 10 | 1016 | 3154293 | ||
BC080721.1 | - | 2 | 683 | |||
D00024.1 | - | 5 | 422 |