Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276684.1 | NP_001263613.1 | 1 | 895 | ||
NM_018790.3 | NP_061260.1 | 1 | 895 | |||
GenBank mRNA | AF162777.1 | - | 1 | 886 | ||
AK157822.1 | - | 1 | 896 | |||
AK170446.1 | - | 1 | 895 | |||
BC023127.1 | - | 1 | 884 | 3498057 |