Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205204.1 | NP_001192133.1 | 14, 15 | 2675 | ||
NM_007495.4 | NP_031521.2 | 14, 15 | 2651 | |||
GenBank mRNA | AB093222.2 | - | 14, 15 | 2237 | ||
AK045561.1 | - | 14, 15 | 2406 | |||
AK138576.1 | - | 14, 15 | 2420 | |||
AK147450.1 | - | 14, 15 | 2421 | |||
AK164117.1 | - | 14, 15 | 2640 | |||
AK165200.1 | - | 14, 15 | 2404 | |||
BC054545.1 | - | 11, 12 | 1425 | |||
BC094666.1 | - | 14, 15 | 2537 | 30545499 | ||
G49230.1 | - | 14, 15 | 2261 | |||
U48797.1 | - | 14, 15 | 2261 |