Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007510.2 | NP_031536.2 | 7, 8 | 601 | ||
GenBank mRNA | AB074758.1 | - | 7, 8 | 603 | ||
AK010603.1 | - | 7, 8 | 617 | |||
AK029146.1 | - | 1388 | ||||
AK139791.1 | - | 7, 8 | 618 | |||
AK146162.1 | - | 7, 8 | 615 | |||
AK149356.1 | - | 7, 8 | 637 | |||
AK167644.1 | - | 7, 8 | 560 | |||
AK169854.1 | - | 7, 8 | 615 | |||
AK208012.1 | - | 1, 2 | 99 | |||
BC003421.1 | - | 7, 8 | 601 | 2598896 | ||
BC055438.1 | - | 7, 8 | 635 | 3995744 |