Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013479.2 | NP_038507.1 | 1 | 468 | ||
GenBank mRNA | AF067660.1 | - | 1 | 465 | ||
AF102501.1 | - | 1 | 439 | |||
AK136172.1 | - | 1 | 469 | |||
AK145385.1 | - | 1 | 336 | |||
AK162127.1 | - | 1 | 336 | |||
BC052690.1 | - | 1 | 467 | 30052580 |