Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077184.1 | NP_001070652.1 | 3 | 452 | ||
NM_009768.2 | NP_033898.1 | 4 | 800 | |||
GenBank mRNA | AK002332.1 | - | 3 | 304 | ||
AK011364.1 | - | 3 | 303 | |||
AK147225.1 | - | 3 | 5126 | |||
AK207023.1 | - | 1 | 51 | |||
AK208329.1 | - | 1 | 52 | |||
AK209680.1 | - | 1 | 52 | |||
AK214501.1 | - | 1 | 52 | |||
AK215038.1 | - | 1 | 52 | |||
AK216955.1 | - | 1 | 52 | |||
AY089967.1 | - | 4 | 658 | |||
BC010270.1 | - | 3 | 295 | 3589236 | ||
D00611.1 | - | 3 | 302 | |||
S63813.1 | - | 3 | 302 | |||
Y16256.1 | - | 3 | 302 | |||
Y16257.1 | - | 2 | 298 | |||
Y16258.1 | - | 2 | 244 |