Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btn1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013483.3 | NP_038511.1 | 8 | 1483 | ||
GenBank mRNA | AK142596.1 | - | 8 | 1390 | ||
AK145168.1 | - | 8 | 1484 | |||
BC011497.1 | - | 8 | 1457 | 3583827 | ||
BC031459.1 | - | 7 | 1381 | 3582574 | ||
S80642.1 | - | 6 | 1209 |