Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009773.3 | NP_033903.2 | 7 | 1019 | ||
GenBank mRNA | AF107296.1 | - | 7 | 974 | ||
AK133909.1 | - | 7 | 960 | |||
BC031577.1 | - | 7 | 970 | 4953223 |