Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009804.2 | NP_033934.2 | 4, 5 | 592 | ||
GenBank mRNA | AK075853.1 | - | 4, 5 | 562 | ||
AK088168.1 | - | 4, 5 | 554 | |||
AK128919.1 | - | 4, 5 | 567 | |||
AK133881.1 | - | 4, 5 | 560 | |||
AK148961.1 | - | 4, 5 | 560 | |||
AK150893.1 | - | 4, 5 | 563 | |||
AK159152.1 | - | 4, 5 | 563 | |||
AK159885.1 | - | 4, 5 | 563 | |||
AK159891.1 | - | 4, 5 | 514 | |||
AK159913.1 | - | 4, 5 | 544 | |||
AK169069.1 | - | 4, 5 | 557 | |||
AK188260.1 | - | 3, 4 | 276 | |||
AK188401.1 | - | 3, 4 | 276 | |||
AY040626.1 | - | 4, 5 | 533 | |||
BC013447.1 | - | 4, 5 | 549 | 4241333 | ||
BC047126.1 | - | 4, 5 | 507 | |||
L25069.1 | - | 4, 5 | 564 | |||
M62897.1 | - | 4, 5 | 477 | |||
X52108.1 | - | 4, 5 | 477 |