Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042072.1 | - | 3 | 361 | ||
AK052396.1 | - | 1 | 2957 | |||
AK133829.1 | - | 3 | 371 | |||
AK170649.1 | - | 2 | 94 | |||
AK197928.1 | - | 2 | 289 | |||
AK199368.1 | - | 3 | 289 | |||
BC034262.1 | - | 3 | 427 | 4460089 | ||
D28492.1 | - | 3 | 350 | |||
Y13085.1 | - | 3 | 429 |